Learn More
Hughes Outdoor Self-Draining Safety Shower with Eye/Face Wash
Product Code. 17732348
Change view
Click to view available options
Material:
Galvanized Steel
Stainless Steel
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
17732348 Stainless Steel Each
17722348 Galvanized Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17732348 Supplier Hughes Supplier No. HSHDSD18SS
  • ONLINE EXCLUSIVE PRICE
    1912.21 €
    1228.76 € / Pack of 1
    Save 683.45 €
    35% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This ANSI compliant self-draining emergency safety shower with open stainless steel eye/face wash, suitable for hot climates.
  • This ANSI compliant self-draining emergency safety shower with open stainless steel eye/face wash, suitable for hot climates
  • The design allows standing water to drain away from the standpipe to ensure the delivery of tepid water
  • Fitted with foot panel as standard
  • The valve is situated directly above the water inlet; the bleed pipe is fitted above this so standing water can drain into the ground preventing stagnant water overheating from solar radiation and removing the risk of further injury to the operator
  • Available with galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Activation Push Lever, Foot Treadle
Material Stainless Steel
Product Type Safety Shower