Learn More
BRAND™ Oval Shape PTFE Magnetic Stirring Bars
Product Code. 10727861 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
10 mm
15 mm
20 mm
25 mm
32 mm
40 mm
50 mm
65 mm
70 mm
Diameter (Metric):
5 mm
6 mm
10 mm
12 mm
15 mm
20 mm
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
10727861 10 mm 5 mm Pack of 10
11782513 25 mm 12 mm Pack of 10
12080922 20 mm 10 mm Pack of 10
11792513 32 mm 15 mm Pack of 10
11864080 40 mm 20 mm Pack of 10
11874080 50 mm 20 mm Pack of 10
10518031 70 mm 20 mm Pack of 10
10029660 15 mm 6 mm Pack of 10
10621862 65 mm 20 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10727861 Supplier BRAND™ Supplier No. 137300
Only null left
Add to basket
Add to basket
PTFE construction. Ideal for vessels with round bottom, such as a round-bottom flask.
  • PTFE - Polytetrafluoroethylene construction
  • Magnetic, Oval shape
  • For vessels with round bottoms
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) For vessels with round-bottom
Diameter (Metric) 5 mm
Length (Metric) 10 mm
Material PTFE
Quantity 10/Pk.
Shape Oval