missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Ovalbumin peptide SIINFEKL (H-2 Kb)
Shop All IBA Lifesciences Products

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Specifications
Specifications
| Purity | >80% |
| Content And Storage | -25°C to -15°C |
| Endotoxin Level | Free |
| Format | Lyophilized Powder |
| Lyophilized | Yes |
| Peptide Tag | H-2 Kb |
| Product Type | Ovalbumin Peptide SIINFEKL |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering