missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Overlay Sheet

Product Code. 12388546
Change view
Click to view available options
For Use With (Equipment):
Fisherbrand GD-4534 Gel Dryer System
Fisherbrand GD-5040 Gel Dryer System
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12388546 Fisherbrand GD-5040 Gel Dryer System Each
11739665 Fisherbrand GD-4534 Gel Dryer System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12388546 Supplier Fisherbrand™ Supplier No. GDSRO

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Accessory for use with vacuum gel dryers.

  • Translucent
  • Silicone rubber construction
  • Allows gels to be visualised during drying process

Specifications

Color Translucent
For Use With (Equipment) Fisherbrand GD-5040 Gel Dryer System
Type Overlay Sheet
Material Silicone Rubber
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.