missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ A40 Light Duty Overshoe with Sole

Product Code. 12695576
Change view
Click to view available options
Size:
Medium/Large
X-Large/2X-Large
Unit Size:
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12695576 X-Large/2X-Large Pack of 200
12685576 Medium/Large Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12695576 Supplier KleenGuard™ Supplier No. 98720
  • ONLINE EXCLUSIVE PRICE
    2336.30 €
    225.51 € / Pack of 200
    Save 2110.79 €
    90% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for keeping footwear clean and covered. KleenGuard™ A40 Light Duty Overshoe with Sole reduces contamination in food processing and light industrial areas.

  • Elasticated overshoe with sole
  • Single-use light duty
  • Ideal for keeping footwear clean and covered
  • Reduce contamination in food processing and light industrial areas
TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Application) Food Industry, Light Industrial
Material Spundbond Polypropylene
Size X-Large/2X-Large
Toe Material Multi-layer Non-woven Barrier Laminate
Waterproof Water Resistant
Product Type Overshoe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.