Learn More
Honeywell™ Overspec Safety Glasses
Product Code. 12657403
Change view
Click to view available options
Lens Tint:
Clear
Gray
Size:
Medium
Large
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint Size Unit Size
12657403 Clear Large Each
12627423 Clear Medium Each
12637423 Clear Medium Each
12677403 Gray Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12657403 Supplier Honeywell™ Supplier No. 1027608
Quantity Price
1 10.74 € / Pack of 1
10 9.58 € / Pack of 1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
OTG protection featuring exceptional peripheral vision/coverage, and a customizable fit.
  • Over-the-glass coverage, allowing workers full protection while wearing prescription eyewear
  • Adjustable and bendable
  • Wire-core temples offer flexibility for a secure, customized fit
  • Ratchetting temples (3 positions) to further customize fit
  • For use in agriculture, manufacturing, and pharmaceutical industries
  • Protection from abrasion, airborne particulates, biohazard, impact
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE: 2, QA: ISO 9001/2000, EC Certificate: C899.1B-D, EC Attestation: C899.3B-D
Lens Coating Scratch-resistant
Lens Tint Clear
For Use With (Application) For use in agriculture, manufacturing, and pharmaceutical industries
Frame Color Blue, Black
Size Large
Lens Material Polycarbonate
Frame Material Nylon