Learn More
Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Standard Comb
Product Code. 12473647
Change view
Click to view available options
Type:
Rapid Load Comb
Teeth Count:
25 (2X)
9 (1X)
Thickness (Metric) Teeth:
1.0 mm
1.5 mm
Unit Size:
Each
Product Code. Type Teeth Count Thickness (Metric) Teeth unitSize
12473647 Rapid Load Comb 25 (2X) 1.0 mm Each
11986916 Rapid Load Comb 25 (2X) 1.5 mm Each
12483647 Rapid Load Comb 9 (1X) 1.0 mm Each
12493647 Rapid Load Comb 9 (1X) 1.5 mm Each
4 options
Product Code. 12473647
Brand: Thermo Scientific™ A1RL25C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs. For use with the Fisherbrand™ Owl™ A1 Large Gel Electrophoresis Systems.
Eliminate comb assembly with this assortment of heavy-duty, one-piece Thermo Scientific™ Owl™ A1 Large Gel Electrophoresis System Combs, for use with Owl A1 Large Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl A1 Large Gel Electrophoresis System
Product Line Owl
Type Rapid Load Comb
Teeth Count 25 (2X)
Thickness (Metric) Teeth 1.0 mm