missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ A1 Large Gel System

Product Code. 11979127
Change view
Click to view available options
Type:
Large Gel Electrophoresis System
Large, w/Buffer Exchange Ports
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11979127 Large Gel Electrophoresis System Each
12403647 Large, w/Buffer Exchange Ports Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11979127 Supplier Thermo Scientific™ Supplier No. A1

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems, which have smaller footprints ideal for detailed RNA/DNA analysis.

Save space with Thermo Scientific™ Owl™ A1 Large Gel Systems. With one of the smallest footprints of any large-format device, these systems are ideal for detailed RNA/DNA analysis.

Features:

  • Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
  • Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
  • Programmable power inverter may be used for field reversal electrophoresis
  • UVT gel tray has gasketed end gates for leakproof seal without tape
  • Built-in leveling ensures flat, uniformly thick gels
  • Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
  • Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
  • Samples may be loaded directly from a 96-well plate, 8 or 12 at a time

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray
  • Gasketed end gates
  • Three combs (12-, 16- and 20-well), 1.5mm thick
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Complete system
Gel Size Mini
Product Line Owl
Type Large Gel Electrophoresis System
Sample Size 8 to 150
Volume (Metric) Buffer 1.6 L
Dimensions (L x W x H) 14.8 x 8.5 x 4.3 in. (37.5 x 21.5 x 11 cm)
Length (English) 14.8 in.
Length (Metric) 37.5 cm
Length (Metric) Gel 25 cm
Width (English) 8.5 in.
Width (Metric) 21.5 cm
Width (Metric) Gel 13 cm
Height (English) 4.3 in.
Height (Metric) 11 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.