missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Owl™ A2 Large Gel Systems
Description
Handle high throughput with consistent, reliable results with Thermo Scientific™Owl™ A2 Large Gel Systems, ideal for detailed RNA/DNA analysis.
Features:
- Gel can be cast in varying lengths to help conserve agarose when fewer samples need to be run
- Optional built-in buffer exchange ports allow for buffer recirculation on extended runs
- UVT gel tray has gasketed end gates for leakproof seal without tape
- Built-in leveling ensures flat, uniformly thick gels
- Comb slots allow two, three or four sample sets of equal lengths to be run on the same gel
- Microwell-format combs provide fast, error-free sample loading with a multichannel pipetter
- Samples may be loaded directly from a 96-well plate, 8 or 12 at a time
Includes:
- Buffer chamber with three-point leveling base
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- UVT gel tray
- Gasketed end gates
- Three combs (16-, 24- and 36-well), 1.5mm thick
Specifications
Specifications
| Certifications/Compliance | CE marked |
| Description | Complete system with buffer exchange ports |
| Product Line | Owl |
| Type | Large, w/Buffer Exchange Ports |
| Sample Size | 8 to 432 (16-, 24- and 36-well combs) |
| Volume (Metric) Buffer | 2.3 L |
| Dimensions (L x W x H) | 14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm) |
| Length (English) | 14.8 in. |
| Length (Metric) | 37.5 cm |
| Length (Metric) Gel | 25 cm |
| Show More |
Product Title
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?