missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ ADJ3 Adjustable System Accessories

Product Code. 11520315 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Gasket
Heat Sink
Size:
45 x 35 cm
65 x 35 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Size unitSize
11520315 Heat Sink 65 x 35 cm Each
11530315 Gasket - Each
11510315 Heat Sink 45 x 35 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11520315 Supplier Thermo Scientific™ Supplier No. ADJ365LM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Owl ADJ3 Adjustable System is the widest adjustable system perfect for sequencing and proteins.

  • The height of the upper buffer chamber (UBC) may be adjusted from 20 cm to 80 cm by sliding the UBC up or down two support poles. The notched or offset glass plate of the gel sandwich presses against a silicone gasket on the UBC for a leakproof seal.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) ADJ1 Giraffe™ Adjustable Gel Electrophoresis System
Type Heat Sink
Size 65 x 35 cm
Thickness (English) 0.18 in.
Thickness (Metric) 0.48 cm
Dimensions (L x W) 25.5 x 13.7 in. (65 x 35 cm)
Length (English) 25.5 in.
Length (Metric) 65 cm
Width (English) 13.7 in.
Width (Metric) 35 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.