missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ Blotting Paper

Product Code. 12424637 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
9 cm
10 cm
12 cm
18 cm
20 cm
35 cm
46 cm
Width (Metric):
9 cm
10 cm
16 cm
20 cm
45 cm
57 cm
Unit Size:
Pack of 100
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Width (Metric) Unit Size
12424637 18 cm 20 cm Pack of 100
12484627 20 cm 20 cm Pack of 100
12494627 35 cm 45 cm Pack of 100
12404637 46 cm 57 cm Pack of 100
12414637 10 cm 10 cm Pack of 100
11932985 9 cm 9 cm Pack of 100
12444637 12 cm 16 cm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12424637 Supplier Thermo Scientific™ Supplier No. FP5
  • ONLINE EXCLUSIVE PRICE
    66.55 €
    63.60 € / Pack of 100
    Save 2.95 €
    4% Off
View all promotions
Estimated Shipment: 01-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with this high-quality blotting paper.

Absorb adequate amounts of buffer for semi-dry electroblotting without the loss of mechanical stability with Thermo Scientific™ Owl™ Blotting Paper, high-quality blotting paper ideal for wicking and blotting applications.

  • High purity, 100% cotton fiber
  • Available in pre-cut mini gel and large format gel sizes
  • Compatible with Owl electroblotting systems

Ordering Information:

  • Sold in packs of 100 sheets
TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 18 cm
Width (Metric) 20 cm
Packaging Type Sheets
Flow Rate 35 mL/min.
For Use With (Equipment) Owl electroblotting systems
Weight 183 g/m2
Format Sheet
Material 100% Cotton Fiber
Dimensions (LxW) 18 cm x 20 cm
Thickness 0.83 mm
Product Line Owl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.