missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts

Product Code. 12464577 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Buffer Chamber and Lid
Casting Slides
Gel Casting Tray
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12464577 Casting Slides Each
12474577 Buffer Chamber and Lid Each
12424587 Gel Casting Tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12464577 Supplier Thermo Scientific™ Supplier No. CGS
  • ONLINE EXCLUSIVE PRICE
    23.95 €
    22.90 € / Pack of 1
    Save 1.05 €
    4% Off
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replace casting slides, casting tray or buffer chamber with Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts.

Replace casting slides, casting tray or buffer chamber with Thermo Scientific™ Owl™ C2-S Micro Electrophoresis System Replacement Parts. Parts are for use with the Owl C2-S Micro Electrophoresis System, a system designed for quick restriction fragment analysis.

  • Glass slides, 5 x 7.5 cm, are used as the gel tray
  • Thin gels can be poured and formed by surface tension directly on the glass slide
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Owl™ C2-S Micro Electrophoresis System
Product Line Owl
Type Casting Slides
Quantity 10 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.