missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ D3-14 Wide Gel Electrophoresis System

Product Code. 11982965
Change view
Click to view available options
Gel Size:
14 x 23 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gel Size Unit Size
11982965 14 x 23 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11982965 Supplier Thermo Scientific™ Supplier No. D314
  • ONLINE EXCLUSIVE PRICE
    2215.00 €
    917.00 € / Pack of 1
    Save 1298.00 €
    58% Off
View all promotions
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Screen PCR products, plasmid preps, restriction mapping and cloning with the Thermo Scientific™ Owl™ D3-14 Wide Gel Electrophoresis System.

Screen PCR products, plasmid preps, restriction mapping and cloning with the Thermo Scientific™ Owl™ D3-14 Wide Gel Electrophoresis System.

Features:
  • Screen 25 to 200 samples on a single agarose gel in less than 30 minutes, producing clear, tight banding patterns with no “smiling.”
  • Using special combs and a multichannel pipetter, samples may be loaded directly from a 96, eight or 12-well plate.
  • Incorporates Owl™ EasyCast™ casting design with external caster—gel tray is fitted with gasket, allowing it to fit snugly into caster and providing a leakproof seal without tape.
  • Sample loading with multichannel pipetter speeds up transfer.
  • Electrode placement allows gel to be run quickly without compromising resolution.
  • Bubble level ensures casting of flat, uniformly thick gels.

Includes:

  • Buffer chamber
  • Thermo Scientific™ SuperSafe™ lid with attached power cords
  • Thermo Scientific™ EasyCast™ gasketed UVT gel tray
  • External caster
  • Four combs (50 micro well, 1.5mm thick)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Gel size L x W: 14 x 23cm; Buffer volume: 800mL; L x W x H: 30 x 27 x 11cm
Gel Size 14 x 23 cm
Product Line Owl
Type Wide
Sample Size 25 to 200
Volume (Metric) Buffer 800 mL
Dimensions (L x W x H) 11.8 x 10.6 x 4.3 in. (30 x 27 x 11 cm)
Length (English) 11.8 in.
Length (Metric) 30 cm
Length (Metric) Gel 14 cm
Width (English) 10.6 in.
Width (English) Gel 6.5
Width (Metric) 27 cm
Width (Metric) Gel 23 cm
Height (English) 4.3 in.
Height (Metric) 11 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.