missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts

Product Code. 12434617
Change view
Click to view available options
Type:
Caster
Gasket
Gel Tray
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12434617 Caster Each
12444617 Gasket Pack of 2
12454617 Gel Tray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12434617 Supplier Thermo Scientific™ Supplier No. D4CST
  • ONLINE EXCLUSIVE PRICE
    247.00 €
    236.00 € / Pack of 1
    Save 11.00 €
    4% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.

Replace your UVT Gel Tray, gaskets and external caster with Thermo Scientific™ Owl™ D4 Horizontal Electrophoresis System Replacement Parts, for use with the Owl D4 System.

  • System design allows you to run one gel tray (51 samples) or stack two gel trays (102 samples) in the buffer chamber while saving valuable bench space.
  • Each of the (2) U.V. Transmissible (UVT) gel trays, 16 x 17 cm (W x L), accommodates up to three combs, allowing the user to run up to three series of samples of equal distances.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Owl™ D4 Electrophoresis System
Product Line Owl
Type Caster
Length (Metric) 17 cm
Width (Metric) 16 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.