missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems

Product Code. 11913015
Change view
Click to view available options
Volume (Metric) Buffer:
300 mL
550 mL
1250 mL
Includes:
Apparatus Only
Complete System
Complete System plus Caster
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Buffer Includes unitSize
11913015 1250 mL Complete System Each
11903015 1250 mL Complete System plus Caster Each
12474677 1250 mL Apparatus Only Each
11913005 300 mL Complete System plus Caster Each
11923005 300 mL Complete System Each
12404707 300 mL Apparatus Only Each
11973005 550 mL Complete System plus Caster Each
11983005 550 mL Complete System Each
11993005 550 mL Apparatus Only Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11913015 Supplier Thermo Scientific™ Supplier No. P10DS2
Estimated Shipment: 11-05-2027
Add to basket
Add to basket
This item is not returnable. View return policy

Quickly and easily cast and run one or two gels in the same device with these Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems.

Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.

Simple, Six-Step Use:

  1. Secure the glass cassettes
  2. Pour the agarose
  3. Plug in the specially designed trough
  4. Cast polyacrylamide solution in the cassettes
  5. Load samples
  6. Run the gel

Features:

  • Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
  • Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
  • Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
  • Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free

Recommended for:

  • SDS electrophoresis
  • The second dimension of two-dimensional (2-D) electrophoresis
  • Agarose electrophoresis
  • Native electrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Product Line Owl
Type Dual-Gel Vertical Electrophoresis System
Gel Dimensions 20 x 20  cm
Includes Complete System
Model P10DS
Volume (Metric) Buffer 1250 mL
Length (English) 12 in.
Length (Metric) Gel 20 cm
Width (English) 6.5 in.
Width (Metric) Gel 20 cm
Height (English) 9 in.
Show More Show Less
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.