Learn More
Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs
Product Code. 11902965 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Double-sided Comb
Prep Comb
Rapid Load Comb
Standard Comb
Wall Comb
Teeth Count:
1
12
16
2
20
24
25 (2X)
8
9 (1X)
Thickness (Metric) Teeth:
1 mm
1.0/1.5 mm
1.5 mm
Unit Size:
Each
Product Code. Type Teeth Count Thickness (Metric) Teeth unitSize
11902965 Double-sided Comb 16 1.0/1.5 mm Each
11952965 Double-sided Comb 12 1.0/1.5 mm Each
11912965 Double-sided Comb 20 1.0/1.5 mm Each
11922965 Double-sided Comb 24 1.0/1.5 mm Each
12474567 Standard Comb 8 1 mm Each
11983509 Standard Comb 8 1.5 mm Each
11962965 Prep Comb 2 1.5 mm Each
12404577 Rapid Load Comb 25 (2X) 1 mm Each
12414577 Rapid Load Comb 25 (2X) 1.5 mm Each
12424577 Rapid Load Comb 9 (1X) 1 mm Each
12434577 Rapid Load Comb 9 (1X) 1.5 mm Each
12444577 Wall Comb 1 1.5 mm Each
12 options
Product Code. 11902965
Brand: Thermo Scientific™ B216
Only null left
Add to basket
Add to basket
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs.
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B2 and B3 Mini Gel Electrophoresis System Combs. These heavy-duty, one-piece combs are for use with the EasyCast™ B2 and B3 Mini Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat. Nos. B2-RL-9C, -9D, -25C and -25D are 8 and 12-channel pipette format (1X) and (2X) combs
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl™ EasyCast B2 and B3 Mini Gel Electrophoresis System
Gel Size Mini
Product Line Owl, EasyCast
Type Double-sided Comb
Teeth Count 16
Thickness (Metric) Teeth 1.0/1.5 mm