missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ P81 Single-Sided Vertical Electrophoresis System, 10 x 10cm gel size, 150mL buffer volume

Product Code. 11903005
Change view
Click to view available options
Length (Metric) Gel:
10 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Gel Unit Size
11903005 10 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11903005 Supplier Thermo Scientific™ Supplier No. P81
  • ONLINE EXCLUSIVE PRICE
    704.00 €
    627.00 € / Pack of 1
    Save 77.00 €
    10% Off
View all promotions
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform quick protein runs with the economical and simple, yet rugged Thermo Scientific™ Owl™ P81 Single-Sided Vertical Electrophoresis System.

Perform quick protein runs with the economical and simple, yet rugged Thermo Scientific™ Owl™ P81 Single-Sided Vertical Electrophoresis System.

Features:
  • Upper buffer chamber extends the length of the gel to distribute heat evenly across the entire gel surface
  • Side clamps press glass plates against silicone gasket to provide uniform pressure and form a leakproof seal resulting in flat, even banding patterns
  • Notched alumina ceramic plate (included) may be used in front of notched glass for cooler, faster runs
Includes:
  • Upper and lower buffer chambers
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • Blank glass plate
  • Notched glass plate
  • Notched alumina plate
  • Two combs (10 well, 0.8mm thick)
  • Spacer set (0.8mm thick)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE marked
Description Gel size: 10 x 10cm; Buffer volume: 150mL; L x W x H: 18 x 8 x 13cm
Product Line Owl
Type Single-Sided Vertical Electrophoresis System
Volume (Metric) Buffer 150 mL
Dimensions (L x W x H) 7.1 x 3.1 x 5.1 in. (18 x 8 x 13 cm)
Length (English) 7.1 in.
Length (Metric) 18 cm
Length (Metric) Gel 10 cm
Width (English) 3.1 in.
Width (Metric) 8 cm
Width (Metric) Gel 10 cm
Height (English) 5.1 in.
Height (Metric) 13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.