missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ P9DS Dual Gel System Side cCamps

Product Code. 11937525
Change view
Click to view available options
Type:
Gasket
Side Clamps
Unit Size:
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
11937525 Gasket Pack of 2
12414707 Side Clamps Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11937525 Supplier Fisherbrand™ Supplier No. P9GK
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Replace blocking plate, buffer chambers, lid or side clamps on your dual gel system with Fisherbrand™ P9DS Dual Gel System Replacement Parts.

Replace blocking plate, buffer chambers, lid or side clamps with Fisherbrand™ P9DS Dual Gel System Replacement Parts, for use with the P9DS Dual Gel System.

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
For Use With (Equipment) Fisherbrand P9DS Gel Electrophoresis System
Type Gasket
Quantity 1 Pair
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.