missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts

Product Code. 12404737
Change view
Click to view available options
Product Type:
Buffer Drain Bottle
Gaskets
Stopcocks
Temperature Indicators
Unit Size:
Each
Pack of 2
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type unitSize
12404737 Gaskets Each
11530495 Buffer Drain Bottle Each
11585235 Stopcocks Pack of 2
12454737 Temperature Indicators Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12404737 Supplier Thermo Scientific™ Supplier No. S4SGK
Add to basket
Add to basket
This item is not returnable. View return policy

Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts.

Achieve optimal manual sequencing with Thermo Scientific™ Owl™ S4S Aluminum-Backed Sequencer System Replacement Parts, for use with the Owl™ S4S Aluminum-Backed Sequencer System.

Choose from these replacement parts for optimal manual sequencing.
TRUSTED_SUSTAINABILITY

Specifications

Description Replacement Gasket
For Use With (Equipment) Owl™ S4S Aluminum-Backed Sequencer System
Product Type Gaskets
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.