missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, 98%, anhydrous

Product Code. 10124043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10124043 5 kg Plastic drum 5kg
10711851 250 g Glass bottle 250g
10701851 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10124043 Supplier Thermo Scientific Acros Supplier No. 186430050
 Restricted Item
Add to basket
This item is not returnable. View return policy

CAS: 144-62-7 | C2H2O4 | 90.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-62-7
Molecular Formula C2H2O4
Molecular Weight (g/mol) 90.034
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
PubChem CID 971
ChEBI CHEBI:16995
IUPAC Name oxalic acid
SMILES C(=O)(C(=O)O)O

Specifications

Melting Point 189.0°C to 191.0°C
Color White
Assay Percent Range 98%
Beilstein 02, 502
Merck Index 15, 7010
Solubility Information Solubility in water: 90g/L (20°C). Other solubilities: very soluble in ethanol, soluble in glycerol, slightly soluble in ether, insoluble in benzene, chloroform and petroleum, ether
Formula Weight 90.04
Percent Purity 98%
Density 1.9000g/mL
Quantity 5 kg
Flash Point >93.4°C
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Crystalline Powder
Specific Gravity 1.9
Chemical Name or Material Oxalic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Oxalic acid, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.