missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Anhydrous, Extra Pure, SLR, Fisher Chemical

Product Code. 10040320 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10040320 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10040320 Supplier Fisher Chemical Supplier No. O/0580/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

pH 1.3
CAS 144-62-7
Melting Point 189°C
Color White
Molecular Formula C2H2O4
MDL Number 2573
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O
IUPAC Name oxalic acid
Molecular Weight (g/mol) 90.034
PubChem CID 971
ChEBI CHEBI:16995
Formula Weight 90.04g/mol
Quantity 500 g
Packaging Plastic powder jar
Physical Form Powder Solid
Vapor Pressure <0.01mmHg at 20°C
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • OXALIC ACID ANHYDROUS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.