missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Concentrate, 0.005M, Solstice

Product Code. 15645260
Change view
Click to view available options
Quantity:
1 Pc.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15645260 1 Pc. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15645260 Supplier Honeywell-Fluka Supplier No. 382551EA
  • ONLINE EXCLUSIVE PRICE
    57.44 €
    54.04 € / Pack of 1
    Save 3.40 €
    5% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

for 1L standard solution, 0.005 M (COOH)2 (0.01N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 144-62-7
Quantity 1 Pc.
Molecular Formula C2H2O4
Linear Formula HOOCCOOH
MDL Number MFCD00002573
UN Number NONH for all modes of transport
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O
IUPAC Name oxalic acid
Molecular Weight (g/mol) 90.034
PubChem CID 971
ChEBI CHEBI:16995
Formula Weight 90.03g/mol
Chemical Name or Material Oxalic acid concentrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.