missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid dihydrate, 99.5+%, ACS reagent

Product Code. 10085830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5kg
Product Code. Quantity Packaging unitSize
10085830 1 kg Plastic bottle 1kg
10300184 250 g Plastic bottle 250g
13408578 5 kg Plastic bottle 5kg
3 options
This item is not returnable. View return policy

Product Code. 10085830

Brand: Thermo Scientific Acros 423150010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
MDL Number MFCD00149102
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Color White
Assay Percent Range 99.5+%
Linear Formula HO2CCO2H·2H2O
UN Number 3261
Beilstein 02, 502
Fieser 01,764; 02,301; 05,481
Merck Index 15, 7010
Formula Weight 126.04
Percent Purity 99.5 to 102.5%
Grade ACS Reagent
Density 4.62
Quantity 1 kg
Infrared Spectrum Authentic
Packaging Plastic bottle
Physical Form Fine Crystalline Powder
Residue after Ignition 0.01% max.
Vapor Pressure 21.5mbar at 50°C
Chemical Name or Material Oxalic acid dihydrate, For analysis ACS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Oxalic acid dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.