missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, ACS Reagent Grade, Ricca Chemical

Product Code. 31751074
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
Poly Wide Mouth Bottle
Unit Size:
125g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31751074 125 g Poly Wide Mouth Bottle 125g
31750187 500 g Poly Wide Mouth Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31751074 Supplier Ricca Chemical Company Supplier No. RDCO0200125B1
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N

Specifications

CAS Max % 102.5
Quantity 125 g
Packaging Poly Wide Mouth Bottle
Residue after Ignition max. 0.01%
SMILES O.O.OC(=O)C(O)=O
IUPAC Name oxalic acid dihydrate
Molecular Weight (g/mol) 126.06
Percent Purity 99.5 to 102.5%
Grade ACS
Chemical Name or Material Oxalic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.