missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid dihydrate, Solstice

Product Code. 16695823
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16695823 500 g 500g
15985920 1 kg 1kg
16605833 5 kg 5kg
16685823 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16695823 Supplier Fluka Supplier No. 33506H500G
  • ONLINE EXCLUSIVE PRICE
    64.00 €
    57.73 € / 500g
    Save 6.27 €
    9% Off
 Restricted Item
Estimated Shipment: 21-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (manganometric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-56-6
PubChem CID 61373

Specifications

Percent Purity 99.5% (min.)
Grade ACS
Quantity 500 g
Loss on Drying 28 - 30 %
Packaging Plastic Bottle
Physical Form Powder
compliance-icons compliance-icons
Product Identifier
  • Oxalic acid dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Oxalic acid dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.