missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Dihydrate, TRC

Product Code. 30012751
Change view
Click to view available options
Quantity:
2.5 g
5 g
25 g
Unit Size:
2.5g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012751 2.5 g 2.5g
30006306 25 g 25g
30011397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012751 Supplier Toronto Research Chemicals Supplier No. TRCO8451332.5G
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C22H24N2O9.2H2O
Molecular Weight (g/mol) 126.0654
Synonym Oxalic Acid Dihydrate, Ethanedioic Acid Dihydrate, Oxaliplatin Imp. A (EP) as Dihydrate, Ethanedioic acid hydrate (1:2)
IUPAC Name oxalic acid;dihydrate
SMILES O.O.OC(=O)C(=O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C2H2O4.2H2O/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);2*1H2
Formula Weight 126.0164
Percent Purity >95%
Chemical Name or Material Oxalic Acid Dihydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.