missing translation for 'onlineSavingsMsg'
Learn More

Oxalic acid, potassium salt dihydrate, 99%, extra pure

Product Code. 10153481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10153481 100 g Glass bottle 100g
10053853 500 g Glass bottle 500g
10609853 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10153481 Supplier Thermo Scientific Acros Supplier No. 352111000
  • ONLINE EXCLUSIVE PRICE
    30.40 €
    28.50 € / 100g
    Save 1.90 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6100-20-5 | C4H3KO8 | 218.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6100-20-5
Molecular Formula C4H3KO8
Molecular Weight (g/mol) 218.16
MDL Number MFCD00150443,MFCD00150443
InChI Key GANDVAJEIJXBQJ-UHFFFAOYSA-M
Synonym potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate
PubChem CID 131698588
IUPAC Name potassium;hydron;oxalate;dihydrate
SMILES [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O

Specifications

Color White to Beige
Loss on Drying 12 % to 16 % (1 g, 105°C)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 7806
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in benzene, ethanol
Formula Weight 254.2
Percent Purity 99%
Grade Extra Pure
Physical Form Crystalline Powder or Solid
Chemical Name or Material Oxalic acid, potassium salt dihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Oxalic acid, potassium salt dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.