missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Solution, 0.1M, Solstice

Product Code. 15684150
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15684150 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15684150 Supplier Honeywell-Fluka Supplier No. 342871L
  • ONLINE EXCLUSIVE PRICE
    152.00 €
    137.38 € / 1 L
    Save 14.62 €
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Volumetric, 0.1 M (COOH)2 (0.2N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

CAS 144-62-7
Quantity 1 L
Molecular Formula C2H2O4
Linear Formula HOOCCOOH
MDL Number MFCD00002573
UN Number NONH for all modes of transport
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O
IUPAC Name oxalic acid
Molecular Weight (g/mol) 90.034
PubChem CID 971
ChEBI CHEBI:16995
Formula Weight 90.03g/mol
Chemical Name or Material Oxalic acid solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.