missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid Solution, Volumetric, 0.05M (COOH)2 (0.1N), Honeywell™ Fluka™

Product Code. 15613550 Shop All Honeywell Fluka Products
Change view
Click to view available options
500mL; Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15613550 500mL; Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15613550 Supplier Honeywell-Fluka Supplier No. 319201500ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 144-62-7
Boiling Point ∼100°C
Color Colorless
Molecular Formula C2H2O4
Linear Formula HOOCCOOH
MDL Number MFCD00002573
UN Number NONH for all modes of transport
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
Solubility Information Soluble in water
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O
IUPAC Name oxalic acid
Molecular Weight (g/mol) 90.034
PubChem CID 971
ChEBI CHEBI:16995
Formula Weight 90.03g/mol
Density 0.99g/mL at 25°C
Quantity 500 mL
Odor Odorless
Packaging Poly bottle
Physical Form Liquid
Vapor Density 4.4 (vs air)
Vapor Pressure 0.01mmHg (20°C)
Chemical Name or Material Oxalic Acid Solution, Volumetric, 0.05M (COOH)2 (0.1N)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.