missing translation for 'onlineSavingsMsg'
Learn More

Oxalic Acid TS, 6.3% (w/v), Ricca

Product Code. 31940599
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31940599 1 L 1 L
31940587 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31940599 Supplier Ricca Chemical Company Supplier No. R54660001A
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Oxalic Acid TS
  • Available in 500 mL and 1 L package sizes in natural poly containers

Chemical Identifiers

Grade ACS

Specifications

CAS 7732-18-5, 6153-56-6
Quantity 1 L
Packaging Natural Poly Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.