missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxaliplatin, 99%

Product Code. 15335058 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 15335058

Brand: Thermo Scientific Chemicals 456131000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61825-94-3 | C8H14N2O4Pt | 397.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 61825-94-3
Melting Point >245.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 2 h)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Molecular Formula C8H14N2O4Pt
MDL Number MFCD00866327
Quantity 100 mg
InChI Key ZROHGHOFXNOHSO-WHMNLBETNA-L
SMILES O=C1O[Pt]OC1=O.N[C@@H]1CCCC[C@H]1N
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine; 1,3-dioxa-2-platinacyclopentane-4,5-dione
Molecular Weight (g/mol) 397.29
Formula Weight 397.29
Percent Purity 99%
Chemical Name or Material Oxaliplatin
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Oxaliplatin
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.