missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxalyl Bromide, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Oxalyl Bromide |
| CAS | 15219-34-8 |
| Molecular Formula | C5 H8 O3 |
| Quantity | 10 g |
| Synonym | Ethanedioyl Bromide, NSC 96957, Oxalyl Dibromide |
| InChI Formula | InChI=1S/C5H8O3/c1-3-4(8-3)5(6)7-2/h3-4H,1-2H3 |
| SMILES | COC(=O)C1OC1C |
| IUPAC Name | methyl 3-methyloxirane-2-carboxylate |
| Molecular Weight (g/mol) | 116.115 |
| Formula Weight | 116.047 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction