missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl Chloride, TRC

Product Code. 30409986
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30409986 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30409986

Brand: Toronto Research Chemicals TRCO84502050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxalyl Chloride
CAS 79-37-8
Molecular Formula C2Cl2O2
Quantity 50 g
Synonym Ethanedioyl Dichloride, Oxalic Acid Chloride, Oxalic Acid Dichloride, Oxalic Dichloride, Oxaloyl Chloride, Oxaloyl Dichloride, Oxalyl Dichloride
InChI Formula InChI=1S/C2Cl2O2/c3-1(5)2(4)6
SMILES ClC(=O)C(=O)Cl
IUPAC Name oxalyl dichloride
Molecular Weight (g/mol) 126.93
Formula Weight 125.9275
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.