missing translation for 'onlineSavingsMsg'
Learn More

Oxamic acid, 98%

Product Code. 10407910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10407910 10 g Glass bottle 10g
10582331 25 g Glass bottle 25g
11965461 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10407910 Supplier Thermo Scientific Acros Supplier No. 129620100
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 471-47-6 | C2H3NO3 | 89.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-47-6
Molecular Formula C2H3NO3
Molecular Weight (g/mol) 89.05
InChI Key SOWBFZRMHSNYGE-UHFFFAOYSA-N
Synonym 2-amino-2-oxoacetic acid, oxamate, acetic acid, aminooxo, oxalamic acid, oxamidic acid, glycine, 2-oxo, oxalic acid monoamide, amino oxo acetic acid, aminooxoacetic acid, oxamate repellent
PubChem CID 974
ChEBI CHEBI:18058
IUPAC Name oxamic acid
SMILES C(=O)(C(=O)O)N

Specifications

Melting Point 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 10 g
Beilstein 02,543
Merck Index 15,7016
Formula Weight 89.05
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Oxamic acid
Show More Show Less
compliance-icons
Product Identifier
  • Oxamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.