missing translation for 'onlineSavingsMsg'
Learn More

Oxamic acid sodium salt, 98%

Product Code. 11407867
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407867 5 g 5g
11417867 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11407867 Supplier Thermo Scientific Alfa Aesar Supplier No. A16532.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 565-73-1 | C2H2NNaO3 | 111.032 g/mol

Sodium oxamate, an inhibitor of LDH-C4, prevented the AR induced by lysophosphatidylcholine (LPC) or NADH. Oxamic acid sodium salt is an lactic acid dehydrogenase. Sperm suspension and cytosol fraction activities were inhibited by sodium oxamate. As cancer cells are often dependent on glycolysis for ATP production, sodium oxamate has implications as an anticancer compound.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium oxamate, an inhibitor of LDH-C4, prevented the AR induced by lysophosphatidylcholine (LPC) or NADH. Oxamic acid sodium salt is an lactic acid dehydrogenase. Sperm suspension and cytosol fraction activities were inhibited by sodium oxamate. As cancer cells are often dependent on glycolysis for ATP production, sodium oxamate has implications as an anticancer compound.

Solubility
Soluble in water (10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565-73-1
Molecular Formula C2H2NNaO3
Molecular Weight (g/mol) 111.032
MDL Number MFCD00044553
InChI Key RQVZIJIQDCGIKI-UHFFFAOYSA-M
Synonym sodium oxamate, oxamic acid sodium salt, sodium 2-amino-2-oxoacetate, oxamic acid, sodium salt, sodium amino oxo acetate, acetic acid, aminooxo-, monosodium salt, oxalic acid monoamide sodium salt, acetic acid, 2-amino-2-oxo-, sodium salt 1:1, oxamic acid sodium, lopac-o-2751
PubChem CID 5242
IUPAC Name sodium;oxamate
SMILES C(=O)(C(=O)[O-])N.[Na+]

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 6538153
Solubility Information Soluble in water (10mg/ml).
Formula Weight 111.03
Percent Purity 98%
Chemical Name or Material Oxamic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.