missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Oxaprozin

Product Code. 16544244 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16544244 5 g 5g
16554244 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16544244 Supplier Thermo Scientific Chemicals Supplier No. 463530050
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21256-18-8
Molecular Formula C18H15NO3
Molecular Weight (g/mol) 293.32
MDL Number MFCD00215977
InChI Key OFPXSFXSNFPTHF-UHFFFAOYSA-N
IUPAC Name 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid
SMILES OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 161.0°C to 165.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohols, very slightly soluble in ether
Formula Weight 293.32
Chemical Name or Material Oxaprozin
compliance-icons
Product Identifier
  • Oxaprozin
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Oxaprozin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.