missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Oxcarbazepine
GREENER_CHOICE

Product Code. 18014642
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18014642 10 mg 10mg
18075712 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18014642 Supplier Tocris Bioscience™ Supplier No. 3864/10
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Anticonvulsant; inhibits Na+ channel activity

Anticonvulsant; protects mice and rats against generalized tonic-clonic seizures induced by electroshock. Thought to act via inhibition of sodium channel activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28721-07-5
Molecular Formula C15H12N2O2

Specifications

Quantity 10 mg
Formula Weight 252.27
Purity >98%
Molecular Weight (g/mol) 252.273
Chemical Name or Material Oxcarbazepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.