missing translation for 'onlineSavingsMsg'
Learn More

Oxetane-3-carboxylic acid, 95%

Product Code. 15404038
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
This item is not returnable. View return policy

Product Code. 15404038

Brand: Thermo Scientific Alfa Aesar H57578.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114012-41-8 | C4H6O3 | 102.089 g/mol

Oxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Slightly soluble in water.

Notes
Store in a cool place. Moisture sensitive. . Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114012-41-8
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD08669505
InChI Key UWOTZNQZPLAURK-UHFFFAOYSA-N
Synonym 3-oxetanecarboxylic acid, 3-oxetanecarboxylicacid, acmc-20ahnr, oxetane-3-carboxylc acd, oxetane-3-carboxylicacid
PubChem CID 19847174
IUPAC Name oxetane-3-carboxylic acid
SMILES C1C(CO1)C(=O)O

Specifications

Density 1.374
Boiling Point 244°C
Flash Point 115°C (239°F)
Quantity 500 mg
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 102.09
Percent Purity 95%
Chemical Name or Material Oxetane-3-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.