missing translation for 'onlineSavingsMsg'
Learn More

Oxidation Reagent 0.02 M Iodine in 78/20/2 THF/Pyridine/Water (v/v), BioSyn™, Honeywell Burdick & Jackson™

Product Code. 16855421
Change view
Click to view available options
Oxidation Reagent 0.02 M Iodine in 78/20/2 THF/Pyridine/Water (v/v), 2500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16855421 Oxidation Reagent 0.02 M Iodine in 78/20/2 THF/Pyridine/Water (v/v), 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16855421 Supplier Honeywell Burdick & Jackson Supplier No. BR6632.5L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

BioSyn™ Honeywell Burdick & Jackson™ reagent solvents for DNA/RNA synthesis

  • Specifically processed and purified to exacting specifications, ensure outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of application
  • All solvents and reagents are filtered (0.2 micron for oxidation reagents and 0.1 micron for all other reagents) to minimize particulate contamination
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Oxidation reagent
Melting Point −108.5°C
CAS 109-99-9
Color Red
CAS Max % 0.78
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Clear liquid
Product Line BioSyn
Boiling Point 66°C

Boiling and melting point temperatures are based off the physical main component.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.