Learn More
Thermo Scientific™ Oxoid™ Amoxycillin Antimicrobial Susceptibility discs
Product Code. 11952962
Change view
Click to view available options
Concentration:
10 μg
2 μg
25 μg
Unit Size:
Pack of 250
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration Unit Size
11952962 10 μg Pack of 250
11933992 25 μg Pack of 250
11953032 2 μg Pack of 250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11952962 Supplier Thermo Scientific™ Supplier No. CT0161B
  • ONLINE EXCLUSIVE PRICE
    36.05 €
    35.18 € / Pack of 250
    Save 0.87 €
    2% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Determine the susceptibility of microorganisms using manual AST methods and a full range of high quality, easy-to-use antimicrobial susceptibility testing discs.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Amoxycillin Antimicrobial Susceptibility discs.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Amoxycillin
Certifications/Compliance Complies with DIN, EUCAST guidelines
Concentration 10 μg
Description Oxoid Amoxycillin Antimicrobial Susceptibility Discs
Antibiotic Code AML
Product Type Antimicrobial Susceptibility Disc
Test Type Antimicrobial Susceptibility
No. per Pack 5 Cartridges/Pk.
Packaging Type 50 Discs/Cartridge
Quantity 50 Discs/Cartridge