missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Anaerobe Basal Broth (Dehydrated)

Product Code. 10556723
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10556723 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10556723 Supplier Thermo Scientific™ Supplier No. CM0957B
  • ONLINE EXCLUSIVE PRICE
    352.00 €
    260.04 € / 500g
    Save 91.96 €
    26% Off
View all promotions
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cultivate anaerobic microorganisms, particularly Bacteroides spp. and other fastidious anaerobes with this medium.

Cultivate anaerobic microorganisms, particularly Bacteroides spp. and other fastidious anaerobes with Thermo Scientific™ Oxoid™ Anaerobe Basal Agar (Dehydrated). Anaerobe Basal Broth is formulated from a range of nutrients which have been selected to optimize the recovery and growth of the majority of anaerobic organisms of clinical importance.

Anaerobe Basal Agar contains peptones, carefully selected to support good growth of anaerobic bacteria and yeast extract as a vitamin source. Starch is present to absorb any toxic metabolites1. Sufficient arginine is added to ensure the growth of Eubacterium lentum2, while haemin and vitamin K are growth factors required by many Bacteroides species3. Haemin is also required by Porphyromonas species. Sodium succinate improves the growth of Prevotella melaninogenica and Bacteroides species4. Sodium pyruvate is added as an energy source for asaccharolytic cocci such as Veillonella. It also acts similarly to catalase and degrades traces of hydrogen peroxide, which may be produced by the action of molecular oxygen on media components5. L-cysteine hydrochloride and dithiothreitol are reducing agents, and cysteine has also been shown to stimulate the growth of some anaerobes6.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description Anaerobe Basal Broth
Form Powder
Product Type Microbiology Media
For Use With (Application) Healthcare
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.