Learn More
Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility disc Dispenser
Product Code. 10210673
Change view
Click to view available options
No. of Places:
6 Oxoid Disc Cartridges
8 Oxoid Disc Cartridges
For Use With (Equipment):
100mm diameter plates
90mm diameter plates
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Places For Use With (Equipment) Unit Size
10210673 6 Oxoid Disc Cartridges 90mm diameter plates Each
10290333 8 Oxoid Disc Cartridges 90mm diameter plates Each
12705610 6 Oxoid Disc Cartridges 100mm diameter plates Each
12657118 8 Oxoid Disc Cartridges 100mm diameter plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10210673 Supplier Thermo Scientific™ Supplier No. ST6090
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods.
Accurately place antimicrobial susceptibility discs onto your media of choice when carrying out manual Antimicrobial Susceptibility Testing methods using a self-tamping Thermo Scientific™ Oxoid™ Antimicrobial Susceptibility disc Dispenser.
One of the simplest and most widely used methods for susceptibility testing, disc diffusion is extremely flexible, offering a wide variety of antibiotics and concentrations which can easily be changed as required.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends onpends on.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance For use only with Oxoid Antimicrobial Susceptibility Discs
Description Oxoid Antimicrobial Susceptibility Disc Dispenser
Product Type Antimicrobial Susceptibility Disc Dispenser
For Use With (Equipment) 90mm diameter plates
No. of Places 6 Oxoid Disc Cartridges
Packaging Type 1 Dispenser/Pk.
For use only with Oxoid™ Antimicrobial Susceptibility discs.