Learn More
Thermo Scientific™ Bacillus Cereus Enterotoxin Reversed Passive Latex Agglutination Kit (BCET – RPLA)
Product Code. 11498299
Change view
Click to view available options
Quantity:
20 Tests/Kit
Unit Size:
20 tests
Product Code. Quantity Unit Size
11498299 20 Tests/Kit 20 tests
1 options
Product Code. 11498299 Supplier Thermo Scientific™ Supplier No. TD0950A
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Rapidly detect Bacillus cereus enterotoxin (diarrhoeal type) in foods and culture filtrates with this reversed passive agglutination assay.
Rapidly detect Bacillus cereus enterotoxin (diarrhoeal type) in foods and culture filtrates with Thermo Scientific™ Oxoid™ Bacillus Cereus Enterotoxin Reversed Passive Latex Agglutination Kit. The technique of reversed passive latex agglutination (RPLA) enables soluble antigens such as bacterial toxins to be detected in an agglutination assay.
The BCET-RPLA test may be used to detect B. cereus enterotoxin in a variety of foods and to give a semi-quantitative result. The test may also be used to demonstrate enterotoxin production by isolates of B. cereus grown in culture.
Bacillus cereus is a common inhabitant of many different environments and can easily contaminate food. If contaminated foods are not cooled sufficiently after cooking and there is an extended time between preparation and consumption of foods, then surviving heat-resistant spores can germinate, enabling the organism to multiply and produce toxins. Under these circumstances, B cereus can cause food poisoning.
Use Bacillus cereus Enterotoxin RPLA test for simple and semi-quantitative detection of staphylococcal enterotoxins in food samples.
  • Simple – easy to implement
  • Visual – easy to read results
In a standard agglutination assay, the soluble antibody reacts with a particulate antigen such as bacterial cells. However, in a reversed agglutination assay the antibody, which is attached to particles, reacts with the soluble antigen. The particles (in this case, latex) do not themselves play a part in the reaction and they are therefore passive. The cross-linking of the latex particles by the specific antigen/antibody reaction results in the visible latex agglutination reaction.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description BCET-RPLA Toxin Detection Kit
Packaging Type 1 Kit
Quantity 20 Tests/Kit