missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ Cronobacter Sakazakii Agar (DFI formulation) (Dehydrated)

Product Code. 10076102
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10076102

Brand: Thermo Scientific™ CM1055B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Differentiate and enumerate Cronobacter sakazakii from infant formula and other food samples with this chromogenic medium.

Differentiate and enumerate Cronobacter sakazakii from infant formula and other food samples with Thermo Scientific™ Brilliance™ Cronobacter Sakazakii Agar (Dehydrated). A chromogenic substrate present in the medium detects a-glucosidase reaction in Cronobacter sakazakii. The enzyme a-glucosidase, present in Chronobacter sakazakii, hydrolyses the substrate producing blue-green colonies on this pale yellow medium.

Cronobacter sakazakii is a Gram-negative rod-shaped bacterium that rarely causes disease in healthy adults. It has been implicated in outbreaks of disease in premature infants (neonates). Research suggests that neonates, or those infants who have other medical conditions, are more susceptible to this infection. Most reported cases of infection are severe, including sepsis (bacteria in the blood, meningitis, or necrotizing enterocolitis (severe intestinal infection).

Use Brilliance Cronobacter Sakazakii Agar for differentiation and enumeration of Cronobacter sakazakii from infant formula and other food samples.

  • Selective: due to the presence of desoxycholate, the medium is selective against most Gram-positive organisms.
  • Easy-to-read: Cronobacter sakazakii produces blue-green colonies on this medium.

Cronobacter sakazakii is an opportunistic pathogen which has been isolated at low levels from powdered infant formulas. The organisms’ high tolerance to desiccation provides a competitive advantage for Cronobacter sakazakii in dry environments, as found in milk powder factories, and thereby increases the risk of post-pasteurization contamination of the finished product1.

The current FDA method1 for the detection of Cronobacter sakazakii is based on yellow pigment production and originated from the pioneering work of Muytjens et al.2. Samples are incubated in water overnight then enriched in EE Broth (CM0317), followed by plating on VBRGA (CM0485) to isolate Enterobacteriaceae. Five colonies are selected and streaked on Tryptone Soya Agar (CM0131), incubated for up to three days and observed for yellow colonies, typical of Cronobacter sakazakii. However, this method does not select for Chronobacter sakazakii and the combined use of EE broth and VRBGA could allow other Enterobacteria to outgrow Chronobacter sakazakii and give false negative results. It is not possible to select for Chronobacter sakazakii colonies from VRBGA plates on the basis of colony morphology as they will appear the same as other Enterobacteria.

TRUSTED_SUSTAINABILITY

Specifications

Description Brilliance E. coli/Coliform Selective Medium
Form Powder
Product Type Agar
Quantity 500 g
Yield For 11.6L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.