missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliant Green Bile Broth 2% (Dehydrated)

Product Code. 10577483
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10577483 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10577483 Supplier Thermo Scientific™ Supplier No. CM0031B

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Detect and confirm the presence of members of coli-aerogenes group in water, wastewater, foods, milk and dairy products with this medium.

Detect and confirm the presence of members of coli-aerogenes group in water, wastewater, foods, milk, and dairy products with Thermo Scientific™ Oxoid™ Brilliant Green Bile Broth 2% (Dehydrated). The medium has been formulated to select for coliform bacteria which are recognised by rapid production of gas through lactose fermentation. Bile salts and brilliant green in the medium inhibit the Gram-positive and Gram-negative bacteria respectively.

Brilliant Green Bile Broth 2% was first described by Durham and Schoenlein1 to selectively recover organisms of the coli-aerogenes group. The concentration of the components of the medium is optimized so that anaerobic lactose fermenters are inhibited without the coliforms being affected.

  • Easy-to-read: coliform bacteria are easy to recognize by rapid production of gas through lactose fermentation.
  • Selects for coliform organisms: bile salts and brilliant green in the medium inhibit nontarget Gram-positive and Gram-negative organisms. The medium is further optimized to prevent the growth of anaerobic lactose fermenters without coliforms being affected.

Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AFNOR, AOAC, BSI, IDF, ISO, NMKL.
Description Brilliant Green Bile 2% Broth
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield for 12.5L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.