missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ C.L.E.D. Medium w/Andrade's Indicator (Dehydrated)

Product Code. 10006012
Click to view available options
Description:
C.L.E.D. Medium w/Andrade's Indicator
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10006012

Brand: Thermo Scientific™ CM0423B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate, enumerate and differentiate urinary pathogens with good colonial differentiation and clear diagnostic characteristics with this medium.

Isolate, enumerate and differentiate urinary pathogens with good colonial differentiation and clear diagnostic characteristics using Thermo Scientific™ Oxoid™ CLED Medium w/Andrade's Indicator (Dehydrated). The medium is a modification of the CLED Medium as described by Mackey and Sandys1. The addition of Andrade's indicator along with bromothymol blue in the medium enhances the appearance of colonies and aids in the identification of microorganisms. The electrolyte deficient nature of CLED Medium prevents the swarming of Proteus species thereby further improving the colonial differentiation.

Use CLED Medium (with Andrade Indicator) to support the growth of the main urinary pathogens for diagnostic urinary bacteriology. Provides easy colony differentiation as Escherichia coli appear as yellow opaque colonies, Proteus form translucent blue colonies, Salmonella appear as flat green colonies while Staphylococcus form deep yellow colonies. Prevents swarming of Proteus species through the electrolyte deficient nature of CLED Medium. Enhances the appearance of colonies and aids identification with the presence of Andrade’s Indicator.

Not all products are available for sale in all territories. Please inquire.  

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Description C.L.E.D. Medium w/Andrade's Indicator
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 13.8L medium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.