missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Campylobacter Growth Supplement

Product Code. 13295409
Change view
Click to view available options
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13295409 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13295409 Supplier Thermo Scientific™ Supplier No. SR0232E
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oxoid Campylobacter Growth Supplement is a liquid supplement for the enhanced growth and aerotolerance of Campylobacter species.

Oxoid Campylobacter Growth Supplement is a liquid supplement for the enhanced growth and aerotolerance of Campylobacter species.

  • For the preparation of Blaser-Wang Medium, Bolton Selective Enrichment Broth, Butzler Selective Medium, and Preston Selective Medium
  • Each vial supplements 500mL of medium
TRUSTED_SUSTAINABILITY

Specifications

Description Campylobacter Growth Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Campylobacter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.