missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cary-Blair Transport Medium (Dehydrated)

Product Code. 10299902
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10299902 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10299902 Supplier Thermo Scientific™ Supplier No. CM0519B
  • ONLINE EXCLUSIVE PRICE
    267.33 €
    251.32 € / 500g
    Save 16.01 €
    5% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Simplify collection, transport, and maintenance of clinical specimens with this transport medium.

Simplify collection, transport, and maintenance of clinical specimens based on the formulation of Cary and Blair1 with Thermo Scientific™ Oxoid™ Cary-Blair Transport Medium (Dehydrated). The low nutrient content of the medium and the use of phosphate as buffering agent prevents bacterial overgrowth by Escherichia coli, Citrobacter freundii and Klebsiella aerogenes.

Ensure optimal sample collection and recovery using the Cary-Blair Transport medium.

  • Prevents bacterial overgrowth by Escherichia coli, Citrobacter freundii and Klebsiella aerogenes.
  • Ensures bacterial survival over long periods of time due to low oxidation reduction potential of the medium2.
  • Particularly suitable in field surveys for Vibrio parahaemolyticus, especially where rectal swabs are to be transported to a central diagnostic laboratory3,4.

For the transport of fastidious anaerobic bacteria, the medium may be prepared as directed and filled into long-narrow screw-capped tubes5. It may also be prepared as a pre-reduced anaerobic sterilized medium (PRAS)6. Methods of producing PRAS media are described by Holdeman and Moore7.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: FDA.
Description Cary-Blair Transport Medium
Form Powder
Product Type Dehydrated Culture Media
Quantity 500 g
Yield For 37.6L medium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.