Learn More
Thermo Scientific™ Oxoid™ CDMN Supplement
Description
Oxoid CDMN Supplement is used for the growth of Clostridium difficile.
- Add to to C. Difficile Agar Base, Part No. CM0601B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | CDMN Selective Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |