Learn More
Thermo Scientific™ China Blue Lactose Agar (Dehydrated)
Description
Enumerate and differentiate bacteria in dairy products with this standard Thermo Scientific™ Oxoid™ China Blue Lactose Agar (Dehydrated). China Blue Lactose Agar, originally formulated by Brandl and Sobeck-skal1, is a standard non-inhibitory solid medium used for the differentiation of lactose fermenters from non-lactose fermenters in milk.
The china blue in the medium serves as a pH indicator to differentiate lactose fermenters from non-lactose fermenters. The medium does not contain any inhibitory substances therefore all the organisms present in milk sample grow luxuriantly on this medium.
Use China Blue Lactose Agar for enumeration and differentiation of bacteria in dairy products. Due to the presence of china blue pH indicator in the medium lactose fermenters are differentiated from non-lactose fermenters.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | China Blue Lactose Agar |
| Form | Powder |
| Product Type | Agar |
| Quantity | 500 g |
| Yield | For 14.3L medium |
For Laboratory Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.